missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541630
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EGAGGGAGTR AGWLATEAQV TQELKGLSGG EERPETEPLL SFVASVSPAD Sequence homology: Human: 100%
TUT1 Polyclonal antibody specifically detects TUT1 in Human samples. It is validated for Western Blot.Spezifikation
TUT1 | |
Polyclonal | |
Unconjugated | |
TUT1 | |
2700038E08Rik; FLJ21850; FLJ22267; FLJ22347; MGC131987; MGC149809; nuclear speckle targeted phosphatidylinositol 4-phosphate 5-kinase type I-alpha regulated-poly(A) polymerase; nuclear speckle-targeted PIPK1A-regulated-poly(A) polymerase; PAP-associated domain-containing 2; PAPD2; poly(A) polymerase associated domain containing 2; RBM21; RNA binding motif protein 21; RNA uridylyltransferase; RNA-binding motif protein 21; RNA-binding protein 21; speckle targeted PIP5K1A-regulated poly(A) polymerase; STARPAP; Star-PAP; terminal uridylyl transferase 1 U6 snRNA-specific; terminal uridylyl transferase 1, U6 snRNA-specific; Tut1; TUTase; TUTase 6; TUTase6; U6 snRNA-specific terminal uridylyltransferase 1; U6 snRNA-specific terminal uridylyltransferase 1; speckle targeted PIP5K1A-regulated poly(A) polymerase; U6 TUTase; U6-TUTase; up-regulated in lung cancer 6; URLC6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64852 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H6E5 | |
TUT1 | |
Synthetic peptide directed towards the middle region of human TUT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |