Learn More
TWEAK Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547065
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) BAFF, rhFas Ligand, rhGITR Ligand, rhOX40 Ligand, rhTRAIL and less than 1% cross-reactivity with rhAPRIL, rhLIGHT, rhTNF-alpha, rhVEGI, and rhTRANCE is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
TWEAK Polyclonal antibody specifically detects TWEAK in Human samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Spezifikation
TWEAK | |
Polyclonal | |
Unconjugated | |
Tnfsf12 | |
APO3 ligand; APO3/DR3 ligand; APO3L; Dr3l; Dr3lg; LOW QUALITY PROTEIN: tumor necrosis factor ligand superfamily member 12; MGC129581; MGC20669; TNF superfamily member 12; TNF-related weak inducer of apoptosis; TNFSF12; TNLG4A; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4A; tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor ligand superfamily member 12, membrane form; Tumor necrosis factor ligand superfamily member 12, secreted form; tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207; zgc:153941 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8742 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O43508 | |
Tnfsf12 | |
E. coli-derived recombinant human TWEAK Arg93-His249 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |