missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYROBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA129027
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-29027 detects TYROBP from human samples.
TYROBP Polyclonal antibody specifically detects TYROBP in Human samples. It is validated for Western Blot.Spezifikation
TYROBP | |
Polyclonal | |
Unconjugated | |
TYROBP | |
DAP12; DNAX-activation protein 12; KARAP; KAR-associated protein; killer cell activating receptor associated protein; killer cell activating receptor-associated protein; killer-activating receptor-associated protein; Ly83; PLOSL; TYRO protein tyrosine kinase binding protein; TYRO protein tyrosine kinase-binding protein; Tyrobp | |
Rabbit | |
Protein G | |
RUO | |
7305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
O43914 | |
TYROBP | |
Recombinant protein - amino acids 28-40 of human TYROBP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |