missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRB Monoclonal Antibody (122.2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA15869
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PTPRB Monoclonal antibody specifically detects PTPRB in Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
PTPRB | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
0 | |
PTPRB | |
Embryonic rat brain proteoglycans. | |
50 μL | |
Primary | |
Rat | |
Antibody | |
IgM |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
122.2 | |
Unconjugated | |
PTPRB | |
3230402H02Rik; C130094E24; HPTPB; HPTP-BETA; protein tyrosine phosphatase, receptor type B; protein tyrosine phosphatase, receptor type, B; protein tyrosine phosphatase, receptor type, beta polypeptide; protein-tyrosine phosphatase beta; PTPB; Ptprb; Ptpz; receptor-type tyrosine-protein phosphatase beta; Rptpb; R-PTP-BETA; vascular endothelial protein tyrosine phosphatase; VEPTP; VE-PTP | |
Mouse | |
Size-exclusion - Dialysis | |
RUO | |
314843 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |