missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYW3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521154
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is A549 cell lysate. PA5-21154 can be used with blocking peptide PEP-1268.
TYW3 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW3 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.Spezifikation
TYW3 | |
Polyclonal | |
Unconjugated | |
TYW3 | |
5230400J09Rik; C1orf171; hypothetical protein BC009514; LOC519731 protein; RGD1559623; tRNA wybutosine-synthesizing protein 3 homolog; tRNA(Phe) 7-((3-amino-3-carboxypropyl)-4-demethylwyosine(37)-N(4))-methyltransferase; tRNA-yW synthesizing protein 3 homolog; tRNA-yW synthesizing protein 3 homolog (S. cerevisiae); tRNA-yW-synthesizing protein 3; Tyw3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127253, 209584 | |
4° C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6IPR3, Q8BSA9 | |
TYW3 | |
A 17 amino acid peptide near the center of human TYW3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |