missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-UBASH3A, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marke: Abnova H00053347-PW1.1Set
Weitere Details : Netto-Gewicht : 0.02000kg
Beinhaltet: Antibody pair for IP: mouse monoclonal anti-UBASH3A (300μg), Antibody pair for WB: rabbit polyclonal anti-UBASH3A (50μL)
Spezifikation
UBASH3A | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
CLIP4/STS-2/TULA | |
Mouse, Rabbit | |
RUO | |
53347 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
UBASH3A | |
UBASH3A | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-UBASH3A (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-UBASH3A (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only