Learn More
YARS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531320
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, U87-MG. Predicted reactivity: Mouse (84%), Rat (83%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
YARS2 Polyclonal antibody specifically detects YARS2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
YARS2 | |
Polyclonal | |
Unconjugated | |
YARS2 | |
2210023C10Rik; CGI-04; EC 6.1.1.1; fi41d10; LOC556702 protein; LOW QUALITY PROTEIN: tyrosine--tRNA ligase, mitochondrial; MLASA2; MT-TYRRS; RGD1311696; tyrosine tRNA ligase 2, mitochondrial; tyrosine--tRNA ligase, mitochondrial; tyrosyl trna synthetase; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2; tyrosyl-tRNA synthetase 2 (mitochondrial); tyrosyl-tRNA synthetase 2, mitochondrial; tyrosyl-tRNA synthetase mitochondrial-like protein; tyrosyl-tRNA synthetase, mitochondrial; tyrRS; wu:fi41d10; Yars2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51067 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.79 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y2Z4 | |
YARS2 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 227 of Human YARS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |