missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534347
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is Hela cell lysate. PA5-34347 can be used with blocking peptide PEP-1390.
YPEL1 Polyclonal antibody specifically detects YPEL1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
YPEL1 | |
Polyclonal | |
Unconjugated | |
YPEL1 | |
0610009L05Rik; 1700016N17Rik; 1700019O22Rik; 4921520K19Rik; 4930511F14Rik; AV118478; cardiac Abnormality/abnormal facies (CATCH22), microdeletion syndrome; Dgl1; DGL-1; DiGeorge syndrome-related protein; FKSG3; mdgl-1; Ppil2; Protein yippee-like 1; yippee like 1; yippee-like 1; yippee-like 1 (Drosophila); YPEL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100360867, 106369, 29799 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O60688, Q9ESC7 | |
YPEL1 | |
An 11 amino acid synthetic peptide near the amino terminus of human YPEL1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |