missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA526957
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine and non-human primate based on sequence homology.
YPEL5 (yippee-like 5) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL5 is expressed at the nucleus and centrosome during interphase; during mitosis, it localizes to the spindle poles, mitotic spindle, and spindle midzone during mitosis. Finally, during cytokinesis, YPEL5 is localized to the midbody. It is associated with the Ran Binding Protein in the Microtubule organizing center (RanBPM) and a related protein RanBP10.Spezifikation
YPEL5 | |
Polyclonal | |
Unconjugated | |
Ypel5 | |
2310076K21Rik; CGI-127; Protein yippee-like 5; yippee like 5; yippee protein homolog; yippee-like 5; yippee-like 5 (Drosophila); Ypel5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
383295, 51646 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P62699, P62700 | |
Ypel5 | |
KLH conjugated synthetic peptide between 92-121 amino acids from the C-terminal region of human YPEL5 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |