missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANXA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15416-1-AP-150UL
This item is not returnable.
View return policy
Description
The annexins include ANXA9 coded protein, which are a family of calcium-dependent phospholipid-binding proteins. Members of the annexin family contain 4 internal repeat domains, each of which includes a type II calcium-binding site. The calcium-binding sites are required for annexins to aggregate and cooperatively bind anionic phospholipids and extracellular matrix proteins. Annexin A9 is a divergent member of the annexin protein family in which all four homologous type II calcium-binding sites in the conserved tetrad core contain amino acid substitutions that ablate their function. However, structural analysis suggests that the conserved putative ion channel formed by the tetrad core is intact.Specifications
| ANXA9 | |
| Polyclonal | |
| Unconjugated | |
| ANXA9 | |
| Annexin 31, Annexin 9, annexin A9, Annexin XXXI, ANX31, ANXA9, Pemphaxin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 71790, 8416 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O76027, Q9JHQ0 | |
| Anxa9 | |
| ANXA9 Fusion Protein Ag7702 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction