Learn More
APOOL Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00 € - 349.00 €
Spezifikation
| Antigen | APOOL |
|---|---|
| Konzentration | 300 μg/mL |
| Anwendungen | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Konjugat | Unconjugated |
| Form | Liquid |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30237995
|
Proteintech
28514-1-AP-150UL |
150 μL |
410.00 € 349.00 € / 150 Mikroliter Sparen 61.00 € 14% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30237600
|
Proteintech
28514-1-AP-20UL |
20 μL |
145.00 € 129.00 € / 20 Mikroliter Sparen 16.00 € 11% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The APOOL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human APOOL. This antibody recognizes human, mouse, rat antigen. The APOOL antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Spezifikation
| APOOL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Liquid | |
| Human, Mouse, Rat | |
| CXorf33; FAM121A; MGC129748; UNQ8193 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 300 μg/mL | |
| Unconjugated | |
| Rabbit | |
| 139322 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |