Learn More
Aquaporin-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Marke: Novus Biologicals NBP1-87679
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Aquaporin-4 Polyclonal specifically detects Aquaporin-4 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Rabbit | |
| 0.1 mL | |
| Rat, Bovine |
| Polyclonal | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CPDVEFKRRFKEAFSKAAQQTKGSYMEVEDNRSQVETDDLILKPGVVHVIDVDRGEEKKGKDQSGEVLSSV | |
| RUO | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |