Learn More
Axl Monoclonal Antibody (MAXL8DS), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
134.00 € - 345.00 €
Spezifikation
Antigen | Axl |
---|---|
Klon | MAXL8DS |
Konzentration | 0.5 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15923314
|
Invitrogen
53108482 |
100 μg |
345.00 €
100 Mikrogramm |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15913314
|
Affymetrix eBioscience
53-1084-80 |
25 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: This MAXL8DS monoclonal antibody recognizes mouse Axl, a member of the TAM family of tyrosine kinase receptors that also includes MerTK and Tyro3. Within hematopoietic compartment the highest expression of Axl can be observed on dendritic cells and macrophages. Stimulation with TLR ligands has been shown to additionally upregulate this expression. This MAXL8DS antibody will work in flow cytometry on both native and fixed/permeabilized cells. Applications Reported: This MAXL8DS antibody has been reported for use in flow cytometric analysis. Applications Tested: This MAXL8DS antibody has been tested by flow cytometric analysis of mouse bone marrow cells cultured with GM-CSF. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
Axl Monoclonal antibody specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.Spezifikation
Axl | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q00993 | |
C57BL/6 mouse Axl-human IgG Fc fusion protein | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
MAXL8DS | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
26362 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |