missing translation for 'onlineSavingsMsg'
Learn More

Azelainsäure, tech. 80 %, Thermo Scientific Chemicals

Catalog Number p-7023574
missing translation for 'orderingAttributeHoverText'
Menge:
500g
2500g
This item is not returnable. View return policy
123-99-9
C9H16O4
188.223
MFCD00004432
BDJRBEYXGGNYIS-UHFFFAOYSA-N
azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
2266
CHEBI:48131
Nicht anedioische Säure
C(CCCC(=O)O)CCCC(=O)O
This item is not returnable. View return policy
123-99-9
C9H16O4
188.223
MFCD00004432
BDJRBEYXGGNYIS-UHFFFAOYSA-N
azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
2266
CHEBI:48131
Nicht anedioische Säure
C(CCCC(=O)O)CCCC(=O)O

It is applied in organic synthesis, lacquers, polyester adhesives, and urethane elastomers. Azelaic acid directly inhibits kallikrein 5 (KLK5), down regulating expression of KLK5, toll-like receptor 2 (TLR 2), and cathelicidin, and suppresses activity of serine proteases. In PUFA-treated fibroblasts, azelaic acid decreases generation of ROS and up regulates expression of antioxidative enzymes, inhibiting cell damage; agonist activity at PPAR? is thought to drive these effects. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Hinweise
Von starken Oxidationsmitteln fernhalten. Halten Sie den Behälter dicht verschlossen und stellen Sie ihn an einen kühlen, trockenen und gut belüfteten Ort.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Azelaic acid
Schmelzpunkt ∼102 °C
Siedepunkt 286 °C (100 mmHg)
Beilstein 1101094
Merck Index 14,905
Löslichkeitsinformationen Soluble in water at 20°C 2.4g/L. Soluble in ethanol (5%),DMSO,methanol (70mg/ml),and alcohol.
Formelmasse 188.22
Reinheit (%) 80%
Güte Eigenschaften
Dichte 1,225
Menge 500g
Flammpunkt 215 °C (419 °F)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'