Learn More
BACH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA562395
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ETMDSETAKM ACPRDQMLPE PISFEAAAIP VAEKEEALLP EPDVPTDTKE SSEKDALTQY PRYKKYQLAC TKNVYNASSH STSGFASTFR EDNSS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
BTB and CNC homolog 2 (BACH2) is a transcriptional regulator that acts as repressor or activator. BACH2 binds to Maf recognition elements (MARE). It induces apoptosis in response to oxidative stress through repression of the antiapoptotic factor HMOX1.Spezifikation
BACH2 | |
Polyclonal | |
Unconjugated | |
BACH2 | |
Bach2; BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB and CNC homology 2; BTB domain and CNC homolog 2; BTBD25; E030004N02Rik; RGD1562865; similar to BTB and CNC homology 1, basic leucine zipper transcription factor 2; transcription regulator protein BACH2; Transcription regulator protein BACH2 (BTB and CNC homolog 2) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60468 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYV9 | |
BACH2 | |
Recombinant protein corresponding to Human BACH2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only