Learn More
BARHL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA552114
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LELSPRSESS SDCSSPASPG RDCLETGTPR PGGASGPGLD SHLQPGQLSA PAQSRTVTSS FLIRDILADC KPLAACAPYS SSGQPAAPEP GGRLAAKAAE DFRDKLDKSG SNASSDSEYK VKEEGDREIS SSRDSP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
BARHL1 (BarH Like Homeobox 1) is a Protein Coding gene. Diseases associated with BARHL1 include Deafness, Autosomal Recessive 27 and Deafness, Autosomal Dominant 16. Gene Ontology (GO) annotations related to this gene include sequence-specific DNA binding and proximal promoter DNA-binding transcription activator activity, RNA polymerase II-specific. An important paralog of this gene is BARHL2.Spezifikation
BARHL1 | |
Polyclonal | |
Unconjugated | |
BARHL1 | |
Bar-class homeodomain protein MBH2; BarH like homeobox 1; BARHL1; Barhl2; BarH-like 1 (Drosophila); barH-like 1 homeobox protein; BarH-like 2; BarH-like homeobox 1; barH-related homeobox protein 1; Bhx1; Dres115; FKSG31; Mbh2; PKCdeltaVIII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54422, 56751 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.35 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P63157, Q9BZE3 | |
BARHL1 | |
Recombinant protein corresponding to Human BARHL1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only