missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-76882-0.025mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Bcl-10 Polyclonal antibody specifically detects Bcl-10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Spezifikation
| Bcl-10 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
| Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human Bcl10. The immunogen is located within the first 50 amino acids of Bcl-10. Amino Acid Squence: APSLTEEDLTEVKKDC | |
| 0.025 mg | |
| Apoptosis, Cancer | |
| 8915 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1-10 μg/mL | |
| AF134395 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur