missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
167.00 € - 507.00 €
Spezifikation
| Antigen | Bcl-10 |
|---|---|
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1-10 μg/mL |
| Anwendungen | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18577430
|
Novus Biologicals
NBP1-76882 |
0.1 mg |
507.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18584944
|
Novus Biologicals
NBP1-76882-0.025mg |
0.025 mg |
167.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Bcl-10 Polyclonal antibody specifically detects Bcl-10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpezifikation
| Bcl-10 | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AF134395 | |
| 8915 | |
| IgG | |
| Peptide affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer | |
| PBS | |
| B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
| Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human Bcl10. The immunogen is located within the first 50 amino acids of Bcl-10. Amino Acid Squence: APSLTEEDLTEVKKDC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts