missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde dimethyl acetal, 97%, BLD Pharm™

Artikelnummer. 31031953
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31031953 25 g
31025006 500 g
31024384 100 g
31020147 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31031953 Supplier BLD Pharm Supplier No. BD11537425g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material Benzaldehyde dimethyl acetal
CAS 1125-88-8
Summenformel C9H12O2
MDL-Nummer MFCD00008491
Menge 25 g
SMILES COC(C1=CC=CC=C1)OC
Molekulargewicht (g/mol) 152.19
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.