missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzene D6 99.96% D, Eurisotop
Description
Benzene D6
- Isotopic Enrichment: 99.96% D
- Quantity: (10) 0.75 mL
- Packaging: Ampule
- Water Content: water <0.01%
CoA and MSDS available at Eurisotop.com
Specifications
Specifications
| Chemischer Name oder Material | Benzene D6 |
| Farbe | Farblos |
| Deuterium (D) | 99.96% D |
| Verpackung | Ampulle |
| Menge | 10 x 0.75 mL |
| CAS | 1076-43-3 |
| Schmelzpunkt | 5.5°C |
| Siedepunkt | 80.1°C |
| Summenformel | C6H6 |
| Synonym | Hexadeuterobenzene, Benzene-D6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?