missing translation for 'onlineSavingsMsg'
Learn More

Benzyl(phenyl)sulfane, 98%, BLD Pharm™

Product Code. 31451207
Encompass_Preferred
Change view
Click to view available options
Menge:
250 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31451207 25 g
31434707 5 g
31451200 250 mg
31444356 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31451207 Supplier BLD Pharm Supplier No. BD11619825g
 Eingeschränkter Artikel
Verfügbar ab: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material Benzyl(phenyl)sulfane
CAS 831-91-4
Summenformel C13H12S
MDL-Nummer MFCD00003066
Menge 25 g
SMILES C1(SCC2=CC=CC=C2)=CC=CC=C1
Molekulargewicht (g/mol) 200.30
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.