Learn More
(+/-)-Benzyloxycarbonyl-Alpha-Phosphonoglycin-Trimethyl-Ester, 97 %, Thermo Scientific Chemicals
Catalog Number 11440697
Click to view available options
Menge:
1 g
5 g
25 g
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
Methyl-2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetat
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
Methyl-2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetat
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
(±)-Z-α-Phosphonoglycin-Trimethylester ist das Ausgangsmaterial für die Synthese von (Z)-Dehydrominsäuren durch die Wittig-Horner-Reaktion mit Aldehyden, die Dehydrominsäuren können stereoselektiv auf Aminosäuren reduziert werden.
Löslichkeit
Löslich in Methanol.
Hinweise
Kühl und trocken in dicht verschlossenen Behältern lagern. Von starken Oxidationsmitteln fernhalten.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester
Schmelzpunkt 76°C to 80°C
Menge 1 g
Löslichkeitsinformationen Soluble in methanol.
Formelmasse 331.27
Reinheit (%) 97%