missing translation for 'onlineSavingsMsg'
Learn More

Bestatin hydrochloride, MedChemExpress

Artikelnummer. 16448126
Encompass_Preferred
Click to view available options
Menge:
10 mg
50 mg
100 mg
Packungsgröße:
10 Milligramm
100 Milligramm
50 Milligramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16448126

Marke: Medchem Express HYB0134A

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer research.

Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer research.

Specifications

Chemischer Name oder Material Bestatin hydrochloride
Physikalische Form Solid
CAS 65391-42-6
Farbe White
Menge 100 mg
Hinweise zur Reinheitsqualität Research
Summenformel C16H25ClN2O4
Zur Verwendung mit (Anwendung) COVID-19-immunoregulation
Synonym Ubenimex hydrochloride
Löslichkeitsinformationen DMSO : 250 mg/mL (724.99 mM; Need ultrasonic) ∣H2O : 100 mg/mL (290.00 mM; Need ultrasonic)
SMILES CC(C)C[C@@H](C(O)=O)NC([C@@H](O)[C@H](N)CC1=CC=CC=C1)=O.Cl
Molekulargewicht (g/mol) 344.83
Formelmasse 344.83
Reinheit (%) 99.17%
Güte Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.