missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA560988
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KEQVANKGEP LALPLNVSEY CVPRGNRR Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 62%.
This gene belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. This gene has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
BEX2 | |
Polyclonal | |
Unconjugated | |
BEX2 | |
BEX1; BEX2; brain expressed X-linked 2; brain-expressed X-linked protein 2; DJ79P11.1; hBex2; protein BEX2; RP1-79P11.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84707 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXY8 | |
BEX2 | |
Recombinant protein corresponding to Human BEX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only