Learn More
BHMT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA560757
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FTFSASEDNM ESKWEDVNAA ACDLAREVAG KGDALVAGGI CQTSIYKYQK DEA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
Homocysteine is a sulfur-containing amino acid that plays a crucial role in methylation reactions. Transfer of the methyl group from betaine to homocysteine creates methionine, which donates the methyl group to methylate DNA, proteins, lipids, and other intracellular metabolites. The protein encoded by this gene is one of two methyl transferases that can catalyze the transfer of the methyl group from betaine to homocysteine. Anomalies in homocysteine metabolism have been implicated in disorders ranging from vascular disease to neural tube birth defects such as spina bifida.Spezifikation
BHMT2 | |
Polyclonal | |
Unconjugated | |
BHMT2 | |
betaine-homocysteine methyltransferase 2; betaine--homocysteine S-methyltransferase 2; betaine-homocysteine S-methyltransferase 2; BHMT2; C81077; D13Ucla2; S-methylmethionine--homocysteine S-methyltransferase BHMT2; SMM-hcy methyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23743, 365972, 64918 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68FT5, Q91WS4, Q9H2M3 | |
BHMT2 | |
Recombinant protein corresponding to Human BHMT2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only