Learn More
ACROBiosystems Biotinylated Cynomolgus Fc gamma RI / CD64 Protein, His, Avitag™ (BLI verified)
Biotinylated BLI verified Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_001270969.1 |
---|---|
Molekulargewicht | 34.8 kDa |
Quelle | Biotinylated Cynomolgus CD64, His,Avitag (FCA-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 11 - Pro 288 (Accession # NP_001270969.1). |
Immunogen | Val 11 - Pro 288 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16427519
|
ACROBiosystems
FCA-C82E8-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16417519
|
ACROBiosystems
FCA-C82E8-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Receptors that recognize the Fc portion of IgG are divided into three groups designated Fc gamma RI, RII, and RIII, also known respectively as CD64, CD32, and CD16. Fc gamma RI binds IgG with high affinity and functions during early immune responses. Fc gamma RII and RIII are low affinity receptors that recognize IgG as aggregates surrounding multivalent antigens during late immune responses.High affinity immunoglobulin gamma Fc receptor I is also known as FCGR1A, FCG1, FCGR1, CD64 and IGFR1, is a type of integral membrane glycoprotein that binds monomeric IgG-type antibodies with high affinity, which belongs to the immunoglobulin superfamily or FCGR1 family. FCGR1A / CD64 contains 3 Ig-like C2-type (immunoglobulin-like) domains. CD64 is constitutively found on only macrophages and monocytes, but treatment of polymorphonuclear leukocytes with cytokines like IFNγ and G-CSF can induce CD64 expression on these cells.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.8 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001270969.1 | |
Biotinylated Cynomolgus CD64, His,Avitag (FCA-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 11 - Pro 288 (Accession # NP_001270969.1). | |
-20°C | |
Fc gamma RI / CD64 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
34.8 kDa | |
Val 11 - Pro 288 | |
FCGR1A,FCG1,FCGR1,IGFR1,CD64,CD64A,FCRI | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |