Learn More
ACROBiosystems Biotinylated Human B7-H4 Protein, Avitag™,His Tag
Recommended for biopanning
Marke: ACROBiosystems B74-H82E2-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
V-set domain-containing T-cell activation inhibitor 1 (VTCN1) is also known as Immune costimulatory protein B7-H4, Protein B7S1, T-cell costimulatory molecule B7x, B7H4, which belongs to the immunoglobulin superfamily and BTN/MOG family. VTCN1 contains two Ig-like V-type (immunoglobulin-like) domains. The expression of VTCN1 is up-regulated by IL6 and IL10 and is inhibited by GM-CSF and IL4 on antigen-presenting cells (APCs). VTCN1 / B7-H4 negatively regulates T-cell-mediated immune response by inhibiting T-cell activation, proliferation, cytokine production and development of cytotoxicity. VTCN1 involved in promoting epithelial cell transformation.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 28 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_078902 | |
25 μg | |
Phe 29 - Ala 258 | |
B7-H4,VTCN1,B7S1,B7h0.5 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
28 kDa | |
Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (B74-H82E2) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # NP_078902). | |
-20°C | |
B7-H4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |