Learn More
ACROBiosystems Biotinylated Human BTLA (31-134) Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems BTA-H82F8-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 40.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
40.5 kDa | |
Biotinylated Human BTLA (31-134), Fc,Avitag (BTA-H82F8) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession # Q7Z6A9). | |
-20°C | |
BTLA | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
25 μg | |
Lys 31 - Thr 134 | |
BTLA,CD272 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |