Learn More
ACROBiosystems Biotinylated Human BTN3A1 / CD277 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems BT1-H82F7-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Butyrophilin subfamily 3 member A1 (BTN3A1) is also known as CD277 and BTF5, which belongs to the immunoglobulin superfamily and contains one B30.2/SPRY domain and two Ig-like V-type (immunoglobulin-like) domains. BTN3A1 plays a role in T-cell activation and in the adaptive immune response. Also, BTN3A1 regulates the proliferation of activated T-cells and the release of cytokines and IFNG by activated T-cells. Furthermore, BTN3A1 mediates the response of T-cells toward infected and transformed cells that are characterized by high levels of phosphorylated metabolites, such as isopentenyl pyrophosphate.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.9 kDa. The protein migrates as 54 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_008979.3 | |
25 μg | |
Gln 30 - Gly 254 | |
BTN3A1,CD277,BTF5 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
52.9 kDa | |
Biotinylated Human BTN3A1, Fc,Avitag (BT1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession # O00481-1). | |
-20°C | |
BTN3A1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |