Learn More
ACROBiosystems Biotinylated Human CD47 Protein, Fc, Avitag™
Biotinylated Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_942088 |
---|---|
Molekulargewicht | 42.2 kDa |
Quelle | MABSol™ Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Accession # NP_942088). |
Immunogen | Gln 19 - Pro 139 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15896277
|
ACROBiosystems
CD7-H82F6-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 06-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15806287
|
ACROBiosystems
CD7-H82F6-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 06-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Leukocyte surface antigen CD47 is also known as Antigenic surface determinant protein OA3, Integrin-associated protein (IAP) and Protein MER6. CD47 contains 1 Ig-like V-type (immunoglobulin-like) domain. CD47 is very broadly distributed on normal adult tissues. CD47 has a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins and plays an important role in memory formation and synaptic plasticity in the hippocampus by similarity. CD47 is the receptor for SIRPA, binding to which prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. CD47 Interaction with SIRPG mediates cell-cell adhesion, enhances superantigen-dependent T-cell-mediated proliferation and costimulates T-cell activation.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 42.2 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_942088 | |
MABSol™ Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Accession # NP_942088). | |
-20°C | |
CD47 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
42.2 kDa | |
Gln 19 - Pro 139 | |
CD47,MER6,IAP,OA3 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |