missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human CD52 Protein, Fc, Avitag™

Biotinylated Protein

Marke:  ACROBiosystems CD2-H82F3-25UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15886157

  • 399.17 € / 25 Mikrogramm
Verfügbar ab: 05-01-2023
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CAMPATH-1 antigen, also known as cluster of differentiation 52 (CD52), is a glycoprotein that in humans is encoded by the CD52 gene.It is widely expressed on the cell surface of immune cells, such as mature lymphocytes, natural killer cells (NK), eosinophils, neutrophils, monocytes/macrophages, and dendritic cells (DCs).ligation of cell surface CD52 molecules may offer costimulatory signals for T-cell activation and proliferation.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 29.7 kDa. As a result of glycosylation, the protein migrates as 38-45 kDa under reducing (R) condition, and 66-100 kDa under no
Spezifikation

Spezifikation

Powder
>95% as determined by SDS-PAGE
Biotinylated Human CD52, Fc,Avitag (CD2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1).
-20°C
CD52
Fc Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
29.7 kDa
25 µg
Gly 25 - Ser 36
CDW52,HE5,CD52,CAMPATH-1 antigen
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote