Learn More
ACROBiosystems Biotinylated Human CTGF / CCN2 Protein, His,Avitag™
Recommended for biopanning
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_001892.1 |
---|---|
Molekulargewicht | 39.2 kDa |
Quelle | Biotinylated Human CTGF, His,Avitag (recommended for biopanning) (CTF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # Q5M8T4-1). |
Immunogen | Gln 27 - Ala 349 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16437459
|
ACROBiosystems
CTF-H82E6-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17106271
|
ACROBiosystems
CTF-H82E6-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Connective Tissue Growth Factor (CTGF), also known as CCN2, is a member of the CCN (CCN1-6) family of modular matricellular proteins. Like other CCN proteins, mature human CTGF consists of IGF-binding protein domain, a vWF-C domain, a TSP-1 domain, and a cysteine knot heparin-binding domain. CTGF promotes proliferation and differentiation of chondrocytes. Mediates heparin- and divalent cation-dependent cell adhesion in many cell types including fibroblasts, myofibroblasts, endothelial and epithelial cells. Enhances fibroblast growth factor-induced DNA synthesis. Analysis of CCN2 function in vivo has focused primarily on its key role as a mediator of excess ECM synthesis in multiple fibrotic diseases.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 39.2 kDa. The protein migrates as 40-42 kDa and 43-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001892.1 | |
Biotinylated Human CTGF, His,Avitag (recommended for biopanning) (CTF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # Q5M8T4-1). | |
-20°C | |
CTGF | |
Human | |
Tris with Potassium glutamate and Arginine, pH7.0 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
39.2 kDa | |
Gln 27 - Ala 349 | |
CCN2,NOV2,HCS24,IGFBP8,IBP-8,IGFBP-8,IGF-binding protein 8 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |