Learn More
ACROBiosystems Biotinylated Human FOLR1 Protein, Fc, Avitag™
Biotinylated Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_057937.1 |
---|---|
Molekulargewicht | 53.3 kDa |
Quelle | Biotinylated Human FOLR1, Fc,Avitag (FO1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1). |
Immunogen | Arg 25 - Met 233 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16447559
|
ACROBiosystems
FO1-H82F9-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16457559
|
ACROBiosystems
FO1-H82F9-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Folate Receptor 1 (FOLR1) is also known as Folate receptor alpha, Folate Binding Protein (FBP), FOLR, and is a member of the folate receptor (FOLR) family. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. Mature FOLR1 is an N-glycosylated protein that is anchored to the cell surface by a GPI linkage. FOLR1 is predominantly expressed on epithelial cells and is dramatically upregulated on many carcinomas. FOLR1 is internalized to the endosomal system where it dissociates from its ligand before recycling to the cell surface. A soluble form of FOLR1 can be proteolytically shed from the cell surface into the serum and breast milk. Defects in FOLR1 are the cause of neurodegeneration due to cerebral folate transport deficiency (NCFTD). NCFTD is an autosomal recessive disorder resulting from brain-specific folate deficiency early in life.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 53.3 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_057937.1 | |
Biotinylated Human FOLR1, Fc,Avitag (FO1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1). | |
-20°C | |
FOLR1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
53.3 kDa | |
Arg 25 - Met 233 | |
FOLR-1,FBP,FOLR | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |