Learn More
ACROBiosystems Biotinylated Human G-CSF R / CD114 Protein, Avitag™,His Tag
Biotinylated Protein
Marke:  ACROBiosystems GCR-H82E4-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15886817
  • 1862.87 € / 200 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The granulocyte colony-stimulating factor receptor (G-CSF-R) also known as CD114 (Cluster of Differentiation 114) is a protein that is encoded by the CSF3R gene. G-CSF-R is a cell-surface receptor for the granulocyte colony-stimulating factor (G-CSF). The G-CSF receptors belongs to a family of cytokine receptors known as the hematopoietin receptor family. Granulocyte colony-stimulating factor receptor has been shown to interact with Grb2, HCK and SHC1. The granulocyte colony-stimulating factor receptor is present on precursor cells in the bone marrow, and, in response to stimulation by G-CSF, initiates cell proliferation and differentiation into mature neutrophilic granulocytes and macrophages.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 69.2 kDa. The protein migrates as 72-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_000751
200 μg
Glu 25 - Pro 621
CSF3R,CD114,GCSFR
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
69.2 kDa
Biotinylated Human G-CSF R, Avitag,His Tag (GCR-H82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751).
-20°C
G-CSF R
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins