Learn More
ACROBiosystems Biotinylated Human IL-17A&IL-17F Protein, Avitag™,His Tag
Biotinylated Protein
Marke: ACROBiosystems ILF-H82W1-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interleukin-17A (IL17A) is also known as cytotoxic T-lymphocyte-associated antigen 8 (CTLA8),which is a proinflammatory cytokine produced by activated T cells. IL17A can regulate the activities of NF-kappaB and mitogen-activated protein kinases. Also,IL17A can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO).Furthermore,IL17A has been found both in glycosylated and nonglycosylated forms. High levels of IL-17 are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
Biotinylated Human IL-17A&IL-17F, Avitag,His Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 34.1 kDa. Subunit IL-17A&IL-17F is fused with an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The reducing (RSpezifikation
NP_002181.1 (IL-17 A) and NP_443104.1 (IL-17 F) | |
25 μg | |
Gly 24 - Ala 155 (IL-17 A) and Arg 31 - Gln 163 (IL-17 F) | |
IL-17 A,Interleukin-17 A,CTLA-8,IL-17,IL-17 F,Interleukin-17 F,Cytokine ML-1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
34.1 kDa | |
Biotinylated Human IL-17 A&IL-17; F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17 A) and Arg 31 - Gln 163 (IL-17 F) (Accession # Q16552-1 (IL-17 A) and Q96PD4-1 (IL-17 F)). | |
-20°C | |
IL-17 A and IL-17 F | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |