Learn More
ACROBiosystems Biotinylated Human LILRA6 / CD85b / ILT8 Protein, His, Avitag™
Biotinylated Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | AGZ61988.1 |
---|---|
Molekulargewicht | 50.0 kDa |
Quelle | Biotinylated Human LILRA6, His,Avitag (LI6-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 447 (Accession # Q6PI73-1). |
Immunogen | Gly 24 - Asn 447 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15857267
|
ACROBiosystems
LI6-H82E2-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 04-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15847267
|
ACROBiosystems
LI6-H82E2-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 04-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The LILRs are a family of receptors that regulate the activities of myelomonocytic cells. LILRB3 and LILRA6 represent a pair of inhibitory/activating receptors with identical extracellular domains and unknown ligands. LILRB3 (ILT5) and LILRA6 (ILT8) are highly polymorphic receptors with similar extracellular domains. LILRA6 can signal through association with an activating adaptor molecule, FcRγ, which bears a cytoplasmic tail with an immunoreceptor tyrosine-based activation motif. Analysis of mRNA expression in the major fractions of PBMCs showed that LILRA6 is primarily expressed in monocytes, and its expression level correlates with copy number of the gene.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.0 kDa. The protein migrates as 56-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AGZ61988.1 | |
Biotinylated Human LILRA6, His,Avitag (LI6-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 447 (Accession # Q6PI73-1). | |
-20°C | |
LILRA6 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
50.0 kDa | |
Gly 24 - Asn 447 | |
LILRA6,CD85b,ILT8,ILT-8 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |