Learn More
ACROBiosystems Biotinylated Human MIF Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems MIF-H82E7-25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Macrophage migration inhibitory factor(MIF) is also known as Glycosylation-inhibiting factor(GIF), L-dopachrome isomerase, L-dopachrome tautomerase, Phenylpyruvate tautomerase. Interacts with CXCR2 extracellular domain, CD74 extracellular domain, COPS5 and BNIPL. The expression of MIF at sites of inflammation suggests a role as mediator in regulating the function of macrophages in host defense. Counteracts the anti-inflammatory activity of glucocorticoids. Has phenylpyruvate tautomerase and dopachrome tautomerase activity (in vitro), but the physiological substrate is not known. It is not clear whether the tautomerase activity has any physiological relevance, and whether it is important for cytokine activity.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.0 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002406.1 | |
25 μg | |
Pro 2 - Ala 115 | |
MIF,GLIF,MMIF,GIF | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
16.0 kDa | |
Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from human 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession # P14174-1). | |
-20°C | |
MIF | |
Human | |
25 mM MES, 500 mM NaCl, pH6.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |