Learn More
ACROBiosystems Biotinylated Human OX40 / TNFRSF4 / CD134 Protein, Avitag™,His Tag
MALS verified
Marke:  ACROBiosystems TN4-H82E4-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887807
  • 399.17 € / 25 Mikrogramm
Verfügbar ab: 23-12-2022
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 22.8 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_003318.1
22.8 kDa
25 µg
Leu 29 - Ala 216
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L
Precision Avi
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Biotinylated Human O x 40, Avitag,His Tag (TN4-H82E4) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318.1).
-20°C
OX40
His Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins