Learn More
ACROBiosystems Biotinylated Human TFPI Protein, His Tag, ultra sensitivity (primary amine labeling)
350.80 € - 1524.16 €
Spezifikation
Zugriffsnummer | NP_006278.1 |
---|---|
Molekulargewicht | 30.0 kDa |
Quelle | MABSol™ Biotinylated Human TFPI, His Tag, primary amine labeling (TFI-H8226) is expressed from human HEK293 cells. It contains AA Asp 29 - Lys 282 (Accession # NP_006278.1). It is the biotinylated form of Human TFPI, His Tag (TFI-H5226). |
Immunogen | Asp 29 - Lys 282 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15867747
|
ACROBiosystems
TFI-H8226-25UG |
25 μg |
350.80 €
25 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15857747
|
ACROBiosystems
TFI-H8226-200UG |
200 μg |
1524.16 €
200 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Tissue factor pathway inhibitor (TFPI) is also known as Extrinsic pathway inhibitor (EPI), Lipoprotein - associated coagulation inhibitor (LACI), is a plasma proteinase inhibitor synthesized by vascular endothelial cells and part of it is associated with glycosaminoglycans of these cells. TFPI is a single-chain polypeptide which can reversibly inhibit Factor Xa (Xa) and Thrombin (Factor IIa). TFPI is a secreted protein with a Nterminal acidic region, three Kunitz (K) domains separated with by two linker regions, and a Cterminal basic region. The first K domain inhibits coagulation factor VIIa complexed to tissue factor (TF); The second K domain inhibits factor Xa; The third K domain binds to heparin; The Cterminal basic region may have several functions. For example, it plays an important role in binding of TFPI to cell surfaces.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.0 kDa. The protein migrates as 41-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006278.1 | |
MABSol™ Biotinylated Human TFPI, His Tag, primary amine labeling (TFI-H8226) is expressed from human HEK293 cells. It contains AA Asp 29 - Lys 282 (Accession # NP_006278.1). It is the biotinylated form of Human TFPI, His Tag (TFI-H5226). | |
-20°C | |
TFPI | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
30.0 kDa | |
Asp 29 - Lys 282 | |
TFPI,LACI,TFPI1,EPI,TFI | |
UltraLys | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |