Learn More
ACROBiosystems Biotinylated Mouse BAFFR / TNFRSF13C Protein, Fc, Avitag™
Biotinylated Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_082351 |
---|---|
Molekulargewicht | 35.1 kDa |
Quelle | Biotinylated Mouse BAFFR, Fc,Avitag (BAR-M82F0) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accession # NP_082351). |
Immunogen | Ser 10 - Ala 71 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
15896007
|
ACROBiosystems
BAR-M82F0-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15886007
|
ACROBiosystems
BAR-M82F0-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Description
BAFF receptor (B-cell activating factor receptor, BAFF-R), also known as tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), is a membrane protein of the TNF receptor superfamily which recognizes BAFF. B-cell activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of BAFF in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 35.1 kDa. As a result of glycosylation, the protein migrates as 45-60 kDa under reducing (R) condition, and 90-116 kDa under noSpecifications
NP_082351 | |
Biotinylated Mouse BAFFR, Fc,Avitag (BAR-M82F0) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accession # NP_082351). | |
-20°C | |
BAFFR | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
35.1 kDa | |
Ser 10 - Ala 71 | |
BAFFR,TNFRSF13C,BROMIX,CD268,CVID4,prolixin,BAFF-R | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |