Learn More
ACROBiosystems Biotinylated Mouse FcRn / FCGRT&B2M Heterodimer Protein, Avitag™,His Tag&Twin-Strep Tag (BLI verified)
Biotinylated Protein
423.38 € - 1862.87 €
Spezifikation
Zugriffsnummer | NP_034319.2 (FCGRT) and NP_033865.2 (B2M) |
---|---|
Molekulargewicht | 34.7 kDa (FCGRT), 14.7 kDa (B2M) |
Quelle | MABSol™ Biotinylated Mouse FCGRT&B2M; Heterodimer Protein, Avitag,His Tag&Twin-Strep; Tag (BLI verified) (FCM-M82W6) is expressed from human HEK293 cells. It contains AA Ser 22 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) (Accession # Q61559-1 (FCGRT) and P01887-1 (B2M)). |
Immunogen | Ser 22 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15876737
|
ACROBiosystems
FCM-M82W6-25UG |
25 μg |
423.38 €
25 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15866737
|
ACROBiosystems
FCM-M82W6-200UG |
200 μg |
1862.87 €
200 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
FCGRT & B2M heterodimer protein (FcRn complex) consist of two subunits: p51 (equivalent to FCGRT), and p14 (equivalent to beta-2-microglobulin), and forms an MHC class I-like heterodimer. Fc fragment of IgG, receptor, transporter, alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.
Biotinylated Mouse FCGRT&B2M Heterodimer Protein, Avitag,His Tag&Twin-Strep Tag (BLI verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 34.7 kDa (FCGRT) and 14.7 kDa (B2M). Subunit FCGRT is fused with an Avi tag (Avitag™) at theSpezifikation
NP_034319.2 (FCGRT) and NP_033865.2 (B2M) | |
MABSol™ Biotinylated Mouse FCGRT&B2M; Heterodimer Protein, Avitag,His Tag&Twin-Strep; Tag (BLI verified) (FCM-M82W6) is expressed from human HEK293 cells. It contains AA Ser 22 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) (Accession # Q61559-1 (FCGRT) and P01887-1 (B2M)). | |
-20°C | |
FcRn (FCGRT and B2M) | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
34.7 kDa (FCGRT), 14.7 kDa (B2M) | |
Ser 22 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) | |
FcRn,FCGRT and B2M | |
Precision Avi | |
His Tag and Strep Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |