missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bolton-Hunter-Reagenz, Thermo Scientific Chemicals
Click to view available options
Menge:
1 g
5 g
Beschreibung
Bolton-Hunter Reagent is used for iodination of proteins that do not contain tyrosine residues, Bolton-Hunter reagent precursor (3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester) is a hydroxysuccinimide ester used as an intermediate for high radioactive radioiodination of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Beschreibung | Powder |
Zur Verwendung mit (Anwendung) | Zur Jodierung von Proteinen, die keine Tyrosinrückstände enthalten |
Molekulargewicht | 263.25g/mol |
Produkttyp | Labeling Reagent |
Menge | 1 g |
Schmelzpunkt | 132° to 133°C |
Inhalt und Lagerung | −30° to −10°C |
Beilstein | 1545011 |
InChI-Schlüssel | KYRUKRFVOACELK-UHFFFAOYSA-N |
SMILES | OC1=CC=C(CCC(=O)ON2C(=O)CCC2=O)C=C1 |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'