Learn More
Bortezomib, MedChemExpress
Beschreibung
Bortezomib (PS-341) is a reversible and selective proteasome inhibitor, and potently inhibits 20S proteasome (Ki=0.6 nM) by targeting a threonine residue. Bortezomib disrupts the cell cycle, induces apoptosis, and inhibits NF-κB. Bortezomib is the first proteasome inhibitor anticancer agent. Anti-cancer activity.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Bortezomib |
| CAS | 179324-69-7 |
| Farbe | White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C19H25BN4O4 |
| Menge | 200 mg |
| Synonym | PS-341 LDP-341 NSC 681239 |
| Löslichkeitsinformationen | Ethanol : 66.67 mg/mL (173.51 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 50 mg/mL (130.13 mM; Need ultrasonic) |
| SMILES | OB(O)[C@H](CC(C)C)NC([C@@H](NC(C1=NC=CN=C1)=O)CC2=CC=CC=C2)=O |
| Molekulargewicht (g/mol) | 384.24 |
| Mehr anzeigen |