Learn More
Bovine Albumin, RIA Grade, Thermo Scientific Chemicals
Used in radioimmuno assay.
Marke: Thermo Scientific J10868.09
Weitere Details : CAS : 9048-46-8 Netto-Gewicht : 1.00000kg
Beschreibung
Suitable for use in radioimmuno assay
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Purity grade: RIA grade
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- MDLNumber: MFCD00130384
- CAS: 9048-46-8
Spezifikation
Albumin | |
10 g | |
4°C | |
Lyophilized |
6.8 to 7.2 | |
Bovine blood | |
Plastic bottle |
RUO – Research Use Only