missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAF [V599E] Recombinant Human Protein, Invitrogen™
BRAF acts upstream of MEK in the MAP kinase pathway.
Marke: Invitrogen PR7304A
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
BRAF is involved in cell division, differentiation, and secretion. The BRAF mutant V599E is oncogenic and found in a variety of cancers.
Spezifikation
NP_004324.1, V599E | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
BRAF V599E | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
67.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
RAF Kinase Family | |
Recombinant | |
Baculovirus |