Learn More
Allied National Brother™ laminiertes Klebeband für PT-550 Beschriftungssystem
Marke: Allied National TZ231
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This tape features our exclusive laminated tape process and our standard adhesive. Perfect for everyday applications. These labels stay on under normal indoor and outdoor use, including hot and cold environments. Best suited for smooth flat surfaces.- Labeling tape for your Brother labelling system.
- Black print on white tape.
- 1-inch width.
- Compatible with PT 330, 350, 520, 540, 580C, 2500PC, 2200, 2400, 530, 550, 9200PC, and 9400 models.
Spezifikation
Weiß | |
Klebeband | |
Laminiertes Band | |
0.5 in. |