missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRP44L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539062
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
BRP44L (brain protein 44-like protein), also known as HSPC040 or CGI-129, is a 109 amino acid mitochondrial protein belonging to the UPF0041 family. The gene that encodes BRP44L maps to human chromosome 6. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson′s disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.Spezifikation
BRP44L | |
Polyclonal | |
Unconjugated | |
MPC1 | |
Mitochondrial pyruvate carrier 1 ,Brain protein 44-like protein,CGI-129, HSPC040, PNAS-115,BRP44L,MPC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171087, 51660, 55951 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y5U8, P63030, P63031 | |
MPC1 | |
A synthetic peptide derived from the internal region of human BRP44L | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |