missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRUNOL5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518805
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternatively spliced transcript variants have been identified in this gene.Spezifikation
BRUNOL5 | |
Polyclonal | |
Unconjugated | |
CELF5 | |
4930565A21Rik; BRUNOL5; BRUNOL-5; bruno-like 5; bruno-like 5 RNA binding protein; bruno-like 5, RNA binding protein; Bruno-like protein 5; CELF5; CELF-5; CUG-BP and ETR-3 like factor 5; CUG-BP- and ETR-3-like factor 5; CUGBP Elav-like family member 5; CUGBP, Elav-like family member 5; RGD1565016; RNA-binding protein BRUNOL-5 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
60680 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8N6W0 | |
CELF5 | |
Synthetic peptide sequence (EPPGGQPDGMKD) corresponding to the internal amino acids of BRUNOL5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |