Learn More
BTBD18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561933
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MPSEVSEVLS VGGRWTPDLE ITSSQPLDGQ EDKLLHVSSL DTPQRSYGDL SPPCSNWVET GLEVSLTTDE LLYPSPKAGK EVSGHSELLG SLPASS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
Specifically required during spermatogenesis to promote expression of piRNA precursors. The piRNA metabolic process mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons, which is essential for the germline integrity. Acts by facilitating transcription elongation at piRNA loci during pachytene.Spezifikation
BTBD18 | |
Polyclonal | |
Unconjugated | |
BTBD18 | |
BTB (POZ) domain containing 18; BTB domain containing 18; BTB/POZ domain-containing protein 18; BTBD18; hCG1730474 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
643376 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B2RXH4 | |
BTBD18 | |
Recombinant protein corresponding to Human BTBD18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only