Learn More
Eurofins DiscoverX BTK Protein, Active
Beschreibung
BTK (Bruton's Tyrosine Kinase) is a member of the Tec family kinases which are important components of both antigen receptor signaling and other cell surface receptors, resulting in regulation of the actin cytoskeleton, adhesion, migration, and transcriptional activation. BTK is found in all cells of the hematopoietic lineage except plasma and T cells, and is required for normal B cell development and signal transduction through cell surface molecules. Mutations in the btk gene result in aberrant B cell development, leading to the X-linked agammaglobulinemia phenotype in humans and to the less severe X-linked immunodeficiency in mice. BTK inhibitors may be of benefit in the treatment of leukemias and lymphomas.
GenBank NM_000061
Spezifikation
Spezifikation
| Zusammensetzung | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekulargewicht | 78.4 kDa |
| Menge | 10 μg |
| Spezies | Human |
| Rekombinant | Recombinant |
| Proteinmarkierung | N-terminal 6His |
| Expressionssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Full-length |
| Reinheits- oder Qualitätsgrad | ≥53% |
| Protein | BTK, active |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.